4'-(Dimethylamino)acetophenone ≥97%

Supplier: Matrix Scientific

084779-1G 084779-10G
102843-356EA 56.92 USD
102843-356 102843-358
4'-(Dimethylamino)acetophenone ≥97%
4'-(Dimethylamino)acetophenone

Hazard: Irritant

Formula: C₁₀H₁₃NO
MDL Number: MFCD00017246
CAS Number: 2124-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR