2-Chloro-4,5-xylenol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4,5-dimethylphenol, 4-Chloro-5-hydroxy-2-xylene, 6-Chloro-3,4-xylenol

084778-1G 084778-10G
102843-352EA 101.54 USD
102843-352 102843-354
2-Chloro-4,5-xylenol ≥97%
2-Chloro-4,5-xylenol

Hazard: Irritant

Formula: C₈H₉ClO
MW: 156.61 g/mol
MDL Number: MFCD00002173
CAS Number: 1124-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR