3-Pyrrolidin-1-ylbenzonitrile ≥97%

Supplier: Matrix Scientific

084774-1G 084774-10G
102843-342EA 184.62 USD
102843-342 102843-344
3-Pyrrolidin-1-ylbenzonitrile ≥97%
3-Pyrrolidin-1-ylbenzonitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₂
MDL Number: MFCD07772828
CAS Number: 175696-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR