1-(3-Cyanophenyl)piperidine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Piperidin-1-ylbenzonitrile
084773-1G
084773-10G
102843-338EA
123.08
USD
102843-338
102843-340
1-(3-Cyanophenyl)piperidine ≥97%
1-(3-Cyanophenyl)piperidine
Hazard: Irritant
Formula:
C₁₂H₁₄N₂ |
MDL Number:
MFCD07368527 CAS Number: 175696-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...