4-(3-Cyanophenyl)morpholine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Morpholinobenzonitrile, 3-(Morpholin-4-yl)benzonitrile

084770-1G 084770-10G
102843-328EA 241.54 USD
102843-328 102843-330
4-(3-Cyanophenyl)morpholine ≥97%
4-(3-Cyanophenyl)morpholine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MW: 188.2300 g/mol
MDL Number: MFCD07368523
CAS Number: 204078-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR