2,3-Dimethoxyphenol ≥97%
Supplier: Matrix Scientific
084764-1G
084764-10G
102843-310EA
29.23
USD
102843-310
102843-312
2,3-Dimethoxyphenol ≥97%
2,3-Dimethoxyphenol
Hazard: Irritant
Formula:
C₈H₁₀O₃ MW: 154.17 g/mol |
MDL Number:
MFCD00008366 CAS Number: 5150-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...