2-(3-Methylphenyl)ethanol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(m-Tolyl)ethanol

084763-1G 084763-10G
102843-306EA 30.77 USD
102843-306 102843-308
2-(3-Methylphenyl)ethanol ≥97%
2-(3-Methylphenyl)ethanol

Hazard: Irritant

Formula: C₉H₁₂O
MW: 136.1900 g/mol
MDL Number: MFCD00002896
CAS Number: 1875-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR