4-Vinylphenyl acetate ≥97%

Supplier: Matrix Scientific

Synonyms: Acetic Acid 4-Vinylphenyl Ester, 4-Acetoxystyrene

084759-10G 084759-50G
102843-294EA 46.15 USD
102843-294 102843-296
4-Vinylphenyl acetate ≥97%
4-Vinylphenyl acetate

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00075734
CAS Number: 2628-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR