2-Chloro-6-fluoro-3-nitrobenzonitrile ≥97%

Supplier: Matrix Scientific

084758-1G 084758-10G
102843-290EA 512.31 USD
102843-290 102843-292
2-Chloro-6-fluoro-3-nitrobenzonitrile ≥97%
2-Chloro-6-fluoro-3-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₂ClFN₂O₂
MDL Number: MFCD00053834
CAS Number: 226419-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR