Benzyl chloromethyl sulfide ≥97%

Supplier: Matrix Scientific

Synonyms: alpha-(Chloromethylthio)toluene

084753-1G 084753-10G
102843-278EA 127.69 USD
102843-278 102843-280
Benzyl chloromethyl sulfide ≥97%
Benzyl chloromethyl sulfide

Hazard: Irritant

Formula: C₈H₉ClS
MW: 172.67 g/mol
MDL Number: MFCD02093436
CAS Number: 3970-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR