3-Amino-4-methylbenzamide ≥97%

Supplier: Matrix Scientific

084739-1G 084739-10G
102843-242EA 101.54 USD
102843-242 102843-244
3-Amino-4-methylbenzamide ≥97%
3-Amino-4-methylbenzamide

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD00035936
CAS Number: 19406-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR