3-Chloro-4'-methoxypropiophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-1-(4-methoxyphenyl)propan-1-one
084736-1G
084736-10G
102843-232EA
298.46
USD
102843-232
102843-234
3-Chloro-4'-methoxypropiophenone ≥97%
3-Chloro-4'-methoxypropiophenone
Hazard: Irritant
Formula:
C₁₀H₁₁ClO₂ MW: 198.65 g/mol |
MDL Number:
MFCD00236602 CAS Number: 35999-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...