1-Methyl-1-phenyl-2-thiourea ≥97%

Supplier: Matrix Scientific

Synonyms: N-Methyl-N-phenylthiourea

084722-1G 084722-10G
102843-194EA 101.54 USD
102843-194 102843-196
1-Methyl-1-phenyl-2-thiourea ≥97%
1-Methyl-1-phenyl-2-thiourea

Hazard: Irritant

Formula: C₈H₁₀N₂S
MDL Number: MFCD00086629
CAS Number: 4104-75-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR