4-Chlorothiobenzamide ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Chlorobenzene-1-carbothioamide

084714-1G 084714-10G
102843-174EA 44.62 USD
102843-174 102843-176
4-Chlorothiobenzamide ≥97%
4-Chlorothiobenzamide

Hazard: Irritant

Formula: C₇H₆ClNS
MW: 171.65 g/mol
MDL Number: MFCD00040956
CAS Number: 2521-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR