3-(4-Methylphenyl)-1-(3-nitrophenyl)prop-2-en-1-one ≥97%

Supplier: Matrix Scientific

084709-1G 084709-10G
102843-162EA 143.08 USD
102843-162 102843-164
3-(4-Methylphenyl)-1-(3-nitrophenyl)prop-2-en-1-one ≥97%
3-(4-Methylphenyl)-1-(3-nitrophenyl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₆H₁₃NO₃
MDL Number: MFCD00047712
CAS Number: 25870-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR