1-(3-Bromopropoxy)-4-nitrobenzene ≥97%

Supplier: Matrix Scientific

084700-1G 084700-10G
102843-142EA 156.92 USD
102843-142 102843-144
1-(3-Bromopropoxy)-4-nitrobenzene ≥97%
1-(3-Bromopropoxy)-4-nitrobenzene

Hazard: Irritant

Formula: C₉H₁₀BrNO₃
MW: 260.09 g/mol
CAS Number: 13094-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR