N-Methylphenethylamine ≥97%

Supplier: Matrix Scientific

084696-1G 084696-10G 084696-50G
102843-128EA 101.54 USD
102843-128 102843-130 102843-132
N-Methylphenethylamine ≥97%
N-Methylphenethylamine

Hazard: Irritant

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00008291
CAS Number: 589-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR