4'-Chlorochalcone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)-3-phenylprop-2-en-1-one, 4'-Chlorobenzylidenacetophenone

084695-1G 084695-10G 084695-50G
102843-122EA 72.31 USD
102843-122 102843-124 102843-126
4'-Chlorochalcone ≥97%
4'-Chlorochalcone

Hazard: Irritant

Formula: C₁₅H₁₁ClO
MDL Number: MFCD00016343
CAS Number: 956-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR