4-(Chloromethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

084693-1G 084693-10G 084693-50G
102843-110EA 101.54 USD
102843-110 102843-112 102843-114
4-(Chloromethyl)benzonitrile ≥97%
4-(Chloromethyl)benzonitrile

Hazard: Irritant

Formula: C₈H₆ClN
MDL Number: MFCD00019761
CAS Number: 874-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR