3-Aminobenzenesulfonamide ≥97%

Supplier: Matrix Scientific

084689-1G 084689-10G 084689-50G
102843-098EA 56.92 USD
102843-098 102843-100 102843-102
3-Aminobenzenesulfonamide ≥97%
3-Aminobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
MDL Number: MFCD00035781
CAS Number: 98-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR