Bis(4-chloro-2-nitrophenyl)disulfide ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4'-Dichloro-2,2'-dinitrodiphenyl Disulphide

084675-1G 084675-10G
102843-064EA 184.62 USD
102843-064 102843-066
Bis(4-chloro-2-nitrophenyl)disulfide ≥97%
Bis(4-chloro-2-nitrophenyl)disulfide

Hazard: Irritant

Formula: C₁₂H₆Cl₂N₂O₄S₂
MW: 377.21 g/mol
MDL Number: MFCD00047692
CAS Number: 2050-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR