(4-Methoxyphenoxy)acetonitrile ≥97%

Supplier: Matrix Scientific

084671-10G 084671-50G
102843-050EA 220 USD
102843-050 102843-052
(4-Methoxyphenoxy)acetonitrile ≥97%
(4-Methoxyphenoxy)acetonitrile

Hazard: Irritant

Formula: C₉H₉NO₂
MW: 163.18
MDL Number: MFCD00017528
CAS Number: 22446-12-4
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR