1-(3-Nitrobenzyl)piperazine ≥97%

Supplier: Matrix Scientific

084668-1G 084668-10G
102843-042EA 184.62 USD
102843-042 102843-044
1-(3-Nitrobenzyl)piperazine ≥97%
1-(3-Nitrobenzyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD02256025
CAS Number: 203047-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR