1-(3-Cyanophenyl)-4-methylpiperazine ≥97%

Supplier: Matrix Scientific

084657-1G 084657-10G
102843-014EA 241.54 USD
102843-014 102843-016
1-(3-Cyanophenyl)-4-methylpiperazine ≥97%
1-(3-Cyanophenyl)-4-methylpiperazine

Hazard: Irritant

Formula: C₁₂H₁₅N₃
MDL Number: MFCD07368541
CAS Number: 204078-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR