4-piperidin-1-ylbenzonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(1-Piperidinyl)benzonitrile

084656-1G 084656-10G
102843-010EA 298.46 USD
102843-010 102843-012
4-piperidin-1-ylbenzonitrile ≥97%
4-piperidin-1-ylbenzonitrile

Hazard: Irritant

Formula: C₁₂H₁₄N₂
MDL Number: MFCD02575368
CAS Number: 1204-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR