Phenyl propargyl ether ≥95%

Supplier: Matrix Scientific

Synonyms: (Prop-2-ynyloxy)benzene, (Prop-2-yn-1-yloxy)benzene

084638-1G 084638-10G 084638-50G
102842-960EA 84.62 USD
102842-960 102842-962 102842-964
Phenyl propargyl ether ≥95%
Phenyl propargyl ether

Hazard: Irritant

Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00048107
CAS Number: 13610-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR