4-Chloro-3-hydrazinobenzotrifluoride ≥97%

Supplier: Matrix Scientific

084633-1G 084633-10G
102842-944EA 156.92 USD
102842-944 102842-946
4-Chloro-3-hydrazinobenzotrifluoride ≥97%
4-Chloro-3-hydrazinobenzotrifluoride

Hazard: Irritant

Formula: C₇H₆ClF₃N₂
MDL Number: MFCD00052685
CAS Number: 1869-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR