Methyl benzimidate hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Benzimidic Acid Methyl Ester Hydrochloride

084632-1G 084632-10G
102842-940EA 86.15 USD
102842-940 102842-942
Methyl benzimidate hydrochloride ≥97%
Methyl benzimidate hydrochloride

Hazard: Irritant

Formula: C₈H₉NO
MW: 171.62 g/mol
MDL Number: MFCD00012575
CAS Number: 5873-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR