3-Benzylamino-1-propanol ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
3-(Benzylamino)propan-1-ol, N-(3-Hydroxypropyl)benzylamine, 3-(Benzylamino)-1-propanol
084629-1G
084629-10G
102842-932EA
21.54
USD
102842-932
102842-934
3-Benzylamino-1-propanol ≥97%
3-Benzylamino-1-propanol
Purity: 97%
CAS: 34579-44-7
MDL Number: MFCD00020622
Molecular Formula: C10H15NO
Molecular Weight: 165,24
Hazard: Irritant
Formula:
C₁₀H₁₅NO MW: 165.24 g/mol |
MDL Number:
MFCD00020622 CAS Number: 4720-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...