3-Bromo-5-(trifluoromethyl)benzonitrile ≥97%
Supplier: Matrix Scientific
084623-1G
084623-10G
102842-916EA
21.54
USD
102842-916
102842-918
3-Bromo-5-(trifluoromethyl)benzonitrile ≥97%
3-Bromo-5-(trifluoromethyl)benzonitrile
Hazard: Irritant
Formula:
C₈H₃BrF₃N |
MDL Number:
MFCD09800708 CAS Number: 691877-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...