1-(4-Chlorophenyl)-3-thien-2-ylprop-2-en-1-one ≥97%

Supplier: Matrix Scientific

084613-1G 084613-10G
102842-888EA 241.54 USD
102842-888 102842-890
1-(4-Chlorophenyl)-3-thien-2-ylprop-2-en-1-one ≥97%
1-(4-Chlorophenyl)-3-thien-2-ylprop-2-en-1-one

Hazard: Irritant

Formula: C₁₃H₉ClOS
MDL Number: MFCD00052383
CAS Number: 6028-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR