3,4-Dimethoxyphenol ≥97%
Supplier: Matrix Scientific
084602-25G
084602-100G
102842-860EA
455.38
USD
102842-860
102842-862
3,4-Dimethoxyphenol ≥97%
3,4-Dimethoxyphenol
Hazard: Irritant
Formula:
C₈H₁₀O₃ |
MDL Number:
MFCD00008390 CAS Number: 2033-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...