2-Propylresorcinol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-n-Propylresorcinol

084592-1G 084592-10G
102842-834EA 376.92 USD
102842-834 102842-836
2-Propylresorcinol ≥97%
2-Propylresorcinol

Hazard: Irritant

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Melting Pt: 101-105°C
MDL Number: MFCD00100568
CAS Number: 13331-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR