Cinnamaldehyde thiosemicarbazone ≥95%

Supplier: Matrix Scientific

084589-1G 084589-10G
102842-824EA 467.69 USD
102842-824 102842-826
Cinnamaldehyde thiosemicarbazone ≥95%
Cinnamaldehyde thiosemicarbazone

Hazard: Irritant

Formula: C₁₀H₁₁N₃S
MDL Number: MFCD00170595
CAS Number: 5351-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR