2-(3-Chlorophenyl)acetohydrazide ≥95%

Supplier: Matrix Scientific

084574-1G 084574-10G
102842-794EA 383.08 USD
102842-794 102842-796
2-(3-Chlorophenyl)acetohydrazide ≥95%
2-(3-Chlorophenyl)acetohydrazide

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD00245851
CAS Number: 66464-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR