1-(2,6-Dichlorophenyl)-2-thiourea ≥98%

Supplier: Matrix Scientific

Synonyms: N-(2,6-Dichlorophenyl)thiourea, 2,6-Dichlorophenylthiourea, 1-(2,6-Dichlorophenyl)thiourea

084569-1G 084569-10G 084569-50G
102842-774EA 56.92 USD
102842-774 102842-776 102842-778
1-(2,6-Dichlorophenyl)-2-thiourea ≥98%
1-(2,6-Dichlorophenyl)-2-thiourea

Hazard: Irritant

Formula: (C₆H₃Cl₂)NC(S)NH₂
MW: 221.11 g/mol
MDL Number: MFCD00041156
CAS Number: 6590-91-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR