4-Fluoro-4'-nitrobenzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Fluorophenyl)(4-nitrophenyl)methanone

084567-1G 084567-10G
102842-766EA 184.62 USD
102842-766 102842-768
4-Fluoro-4'-nitrobenzophenone ≥97%
4-Fluoro-4'-nitrobenzophenone

Hazard: Irritant

Formula: C₁₃H₈FNO₃
MW: 245.2059 g/mol
MDL Number: MFCD00053428
CAS Number: 2195-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR