(4-tert-Butylphenyl)methanethiol ≥97%

Supplier: Matrix Scientific

084565-10G 084565-50G
102842-760EA 2692.31 USD
102842-760 102842-762
(4-tert-Butylphenyl)methanethiol ≥97%
(4-tert-Butylphenyl)methanethiol

Hazard: Irritant

Formula: C₁₁H₁₆S
MW: 180.31 g/mol
MDL Number: MFCD00068220
CAS Number: 49543-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR