3-(3-Fluorophenyl)propiononitrile ≥97%
Supplier: Matrix Scientific
084563-1G
084563-10G
102842-754EA
467.69
USD
102842-754
102842-756
3-(3-Fluorophenyl)propiononitrile ≥97%
3-(3-Fluorophenyl)propiononitrile
Hazard: Irritant
Formula:
C₉H₈FN |
MDL Number:
MFCD08235267 CAS Number: 25468-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...