4-(3-Nitrophenyl)morpholine ≥97%

Supplier: Matrix Scientific

084558-1G 084558-10G
102842-742EA 255.38 USD
102842-742 102842-744
4-(3-Nitrophenyl)morpholine ≥97%
4-(3-Nitrophenyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD03195994
CAS Number: 116922-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR