(4-Aminophenyl)acetonitrile hydrochloride ≥97%

Supplier: Matrix Scientific

084557-1G 084557-10G 084557-50G
102842-736EA 101.54 USD
102842-736 102842-738 102842-740
(4-Aminophenyl)acetonitrile hydrochloride ≥97%
(4-Aminophenyl)acetonitrile hydrochloride

Hazard: Irritant

Formula: C₈H₉ClN₂
MDL Number: MFCD00017650
CAS Number: 3457-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR