4-Bromo-6-chloro-o-cresol ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-2-chloro-6-methylphenol
084543-1G
084543-50G
102842-698EA
56.92
USD
102842-698
102842-702
4-Bromo-6-chloro-o-cresol ≥97%
4-Bromo-6-chloro-o-cresol
Hazard: Irritant
Formula:
C₇H₆BrClO MW: 221.4800 g/mol |
MDL Number:
MFCD00002167 CAS Number: 7530-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...