4-Brom-2-(trifluoromethoxy)phenyl isothiocyanate ≥97%
Supplier: Matrix Scientific
084541-1G
084541-10G
102842-692EA
101.54
USD
102842-692
102842-694
4-Brom-2-(trifluoromethoxy)phenyl isothiocyanate ≥97%
4-Brom-2-(trifluoromethoxy)phenyl isothiocyanate
Hazard: Irritant
Formula:
C₅H₅BrF₃NOS MW: 264.07 g/mol |
CAS Number:
238742-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...