(2-Methoxyphenoxy)acetonitrile ≥97%
Supplier: Matrix Scientific
084532-1G
084532-10G
102842-668EA
113.85
USD
102842-668
102842-670
(2-Methoxyphenoxy)acetonitrile ≥97%
(2-Methoxyphenoxy)acetonitrile
Hazard: Irritant
Formula:
C₉H₉NO₂ |
CAS Number:
6781-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...