[(4-Fluorophenyl)sulfonyl]acetonitrile ≥97%

Supplier: Matrix Scientific

084523-1G 084523-10G
102842-644EA 127.69 USD
102842-644 102842-646
[(4-Fluorophenyl)sulfonyl]acetonitrile ≥97%
[(4-Fluorophenyl)sulfonyl]acetonitrile

Hazard: Irritant

Formula: C₈H₆FNO₂S
MDL Number: MFCD00179339
CAS Number: 32083-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR