3-Fluorobenzenesulfonamide ≥97%
Supplier: Matrix Scientific
084517-1G
084517-10G
102842-630EA
127.69
USD
102842-630
102842-632
3-Fluorobenzenesulfonamide ≥97%
3-Fluorobenzenesulphonamide
Hazard: Irritant
Formula:
C₆H₆FNO₂S MW: 175.18 g/mol |
MDL Number:
MFCD00042284 CAS Number: 1524-40-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...