(4-Aminophenyl)acetonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(4-Aminophenyl)acetonitrile, 4-Aminobenzyl cyanide
084512-1G
084512-10G
084512-50G
102842-614EA
72.31
USD
102842-614
102842-616
102842-618
(4-Aminophenyl)acetonitrile ≥97%
(4-Aminophenyl)acetonitrile
Hazard: Irritant
Formula:
C₈H₈N₂ MW: 132.17 g/mol |
MDL Number:
MFCD00007912 CAS Number: 3544-25-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...