Cyclohexyl(phenyl)methanol ≥97%
Supplier: Matrix Scientific
084494-1G
084494-10G
102842-572EA
255.38
USD
102842-572
102842-574
Cyclohexyl(phenyl)methanol ≥97%
Cyclohexyl(phenyl)methanol
Hazard: Irritant
Formula:
C₁₃H₁₈O |
MDL Number:
MFCD00019357 CAS Number: 945-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...