N-(4-Acetyl-3-hydroxyphenyl)acetamide ≥97%

Supplier: Matrix Scientific

084493-1G 084493-10G
102842-568EA 426.15 USD
102842-568 102842-570
N-(4-Acetyl-3-hydroxyphenyl)acetamide ≥97%
N-(4-Acetyl-3-hydroxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD00026134
CAS Number: 40547-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR