Bis(3-nitrophenyl) disulfide ≥97%

Supplier: Matrix Scientific

084492-1G 084492-10G 084492-50G
102842-562EA 56.92 USD
102842-562 102842-564 102842-566
Bis(3-nitrophenyl) disulfide ≥97%
Bis(3-nitrophenyl)disulphide

Hazard: Irritant

Formula: C₁₂H₈N₂O₄S₂
MDL Number: MFCD00007246
CAS Number: 537-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR